Gene Transcripts

Gene Symbol
Zcchc9
Entrez Gene ID
309986
Gene Name
zinc finger, CCHC domain containing 9
Gene Aliases
Chromosome Location
Chr. 2: 20909764 - 20919848 on Build Rnor_6.0
UniGene ID
Rn.94285
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013156.1 NP_001013174.1 5 961
XM_006231841.2 XP_006231903.1 6 1063
GenBank mRNA BC083695.1 - 5 961

View Full Product Details