Gene Transcripts

Gene Symbol
Tnfaip8l2
Entrez Gene ID
310663
Gene Name
tumor necrosis factor, alpha-induced protein 8-like 2
Gene Aliases
RGD1310519
Chromosome Location
Chr. 2: 196309789 - 196323881 on Build Rnor_6.0
UniGene ID
Rn.35002
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014039.1 NP_001014061.1 3 538
XM_008761303.1 XP_008759525.1 2 1291
GenBank mRNA BC079019.1 - 3 538
FQ219780.1 - 2 249

View Full Product Details