Gene Transcripts

Gene Symbol
Noc4l
Entrez Gene ID
360828
Gene Name
nucleolar complex associated 4 homolog (S. cerevisiae)
Gene Aliases
RGD1310661
Chromosome Location
Chr. 12: 52072190 - 52076728 on Build Rnor_6.0
UniGene ID
Rn.8908
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014129.1 NP_001014151.1 15 1717
GenBank mRNA BC088275.1 - 15 1717

View Full Product Details