Gene Transcripts

Gene Symbol
Cox7a1
Entrez Gene ID
12865
Gene Name
cytochrome c oxidase subunit VIIa 1
Gene Aliases
COX7A, COX7AH, COX7AM
Chromosome Location
Chr. 7: 30184171 - 30186030 on Build GRCm38
UniGene ID
Mm.423030
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009944.3 NP_034074.1 2 127
GenBank mRNA AF037370.1 - 2 90
AK079950.1 - 2 128
BC060974.1 - 2 105 6543330

View Full Product Details