Gene Transcripts

Gene Symbol
Olfr32
Entrez Gene ID
18331
Gene Name
olfactory receptor 32
Gene Aliases
MOR227-7P, MOR227-9_p, MTPCR05
Chromosome Location
Chr. 2: 90138081 - 90268639 on Build GRCm38
UniGene ID
Mm.246535
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010980.2 NP_035110.1 5 682
GenBank mRNA BC119404.1 - 1 457
BC120653.1 - 1 457 40129576
X89678.1 - 1 34

View Full Product Details