Gene Transcripts

Gene Symbol
Prss41
Entrez Gene ID
71003
Gene Name
protease, serine 41
Gene Aliases
4931440B09Rik, Tessp-1, Tessp1
Chromosome Location
Chr. 17: 23836768 - 23844374 on Build GRCm38
UniGene ID
Mm.328839
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027644.1 NP_081920.1 4 197
GenBank mRNA AB049453.1 - 4 197
AK016509.1 - 4 175
AK049297.1 - 4 214

View Full Product Details