Gene Transcripts

Gene Symbol
Naaa
Entrez Gene ID
497009
Gene Name
N-acylethanolamine acid amidase
Gene Aliases
Asahl
Chromosome Location
Chr. 14: 17283262 - 17302364 on Build Rnor_6.0
UniGene ID
Rn.101943
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001010967.2 NP_001010967.1 4 555
GenBank mRNA AB162193.1 - 4 553
BC105771.1 - 4 555 7930474

View Full Product Details