Gene Transcripts

Gene Symbol
Tmem81
Entrez Gene ID
498229
Gene Name
transmembrane protein 81
Gene Aliases
Chromosome Location
Chr. 13: 49277722 - 49280153 on Build Rnor_6.0
UniGene ID
Rn.41478
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001017490.1 NP_001017490.1 2 822
GenBank mRNA BC079284.1 - 2 822

View Full Product Details