Gene Transcripts

Gene Symbol
Gga2
Entrez Gene ID
74105
Gene Name
golgi associated, gamma adaptin ear containing, ARF binding protein 2
Gene Aliases
1200007E24Rik, mKIAA1080
Chromosome Location
Chr. 7: 121986722 - 122021198 on Build GRCm38
UniGene ID
Mm.29619
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028758.2 NP_083034.1 17 3196
GenBank mRNA AK143691.1 - 17 3184
BC062933.1 - 17 3165 30356060

View Full Product Details