Gene Transcripts

Gene Symbol
Gga2
Entrez Gene ID
74105
Gene Name
golgi associated, gamma adaptin ear containing, ARF binding protein 2
Gene Aliases
1200007E24Rik, mKIAA1080
Chromosome Location
Chr. 7: 121986722 - 122021198 on Build GRCm38
UniGene ID
Mm.29619
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028758.2 NP_083034.1 10 1019
GenBank mRNA AK004632.1 - 10 991
AK034612.1 - 7 662
AK051911.1 - 1 90
AK129280.1 - 10 978
AK143691.1 - 10 1007
AK149798.1 - 10 1009
AK154904.1 - 10 994
AK159953.1 - 10 1009
AK196854.1 - 1 23
BC057377.1 - 3 522 5698559
BC062933.1 - 10 988 30356060

View Full Product Details