Gene Transcripts
- Gene Symbol
- APPL2
- Entrez Gene ID
- 55198
- Gene Name
- adaptor protein, phosphotyrosine interacting with PH domain and leucine zipper 2
- Gene Aliases
- DIP13B
- Chromosome Location
- Chr. 12: 105173297 - 105236230 on Build GRCh38
- UniGene ID
- Hs.506603
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001251904.1 |
NP_001238833.1
|
5, 6 |
589 |
|
NM_001251905.1 |
NP_001238834.1
|
5, 6 |
632 |
|
NM_018171.3 |
NP_060641.2
|
5, 6 |
589 |
|
XM_006719472.1 |
XP_006719535.1
|
5, 6 |
508 |
|
XM_011538530.2 |
XP_011536832.1
|
5, 6 |
2041 |
|
XM_011538531.2 |
XP_011536833.1
|
5, 6 |
1704 |
|
XM_011538532.2 |
XP_011536834.1
|
5, 6 |
3030 |
|
XM_017019551.1 |
XP_016875040.1
|
5, 6 |
994 |
|
XM_017019552.1 |
XP_016875041.1
|
5, 6 |
1704 |
|
XM_017019553.1 |
XP_016875042.1
|
6, 7 |
3302 |
|
XM_017019554.1 |
XP_016875043.1
|
5, 6 |
508 |
|
GenBank mRNA |
AK001521.2 |
- |
5, 6 |
409 |
|
AK290562.1 |
- |
5, 6 |
442 |
|
AK293783.1 |
- |
5, 6 |
381 |
|
AK296610.1 |
- |
5, 6 |
388 |
|
AK297100.1 |
- |
5, 6 |
442 |
|
AK310377.1 |
- |
5, 6 |
377 |
|
AY113704.1 |
- |
5, 6 |
589 |
|
BC033731.1 |
- |
5, 6 |
493 |
5168590 |
View Full Product Details