Gene Transcripts

Gene Symbol
MAST4
Entrez Gene ID
375449
Gene Name
microtubule associated serine/threonine kinase family member 4
Gene Aliases
Chromosome Location
Chr. 5: 66596348 - 67169595 on Build GRCh38
UniGene ID
Hs.595458
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164664.1 NP_001158136.1 6 1127
NM_001290226.1 NP_001277155.1 3 570
NM_001290227.1 NP_001277156.1 4 351
NM_001297651.1 NP_001284580.1 3 611
NM_015183.2 NP_055998.1 5 320
XM_006714606.3 XP_006714669.1 4 573
XM_006714610.2 XP_006714673.1 3 560
XM_011543382.2 XP_011541684.1 8 1219
XM_011543384.2 XP_011541686.1 3 2278
XM_011543385.2 XP_011541687.1 3 3398
XM_011543386.2 XP_011541688.1 3 229
XM_017009447.1 XP_016864936.1 9 1300
XM_017009448.1 XP_016864937.1 7 1183
XM_017009449.1 XP_016864938.1 4 618
XM_017009450.1 XP_016864939.1 4 312
XM_017009451.1 XP_016864940.1 4 312
XM_017009452.1 XP_016864941.1 4 3627
XM_017009453.1 XP_016864942.1 3 10442
GenBank mRNA AK057601.1 - 3 564
AK131421.1 - 4 652
AK296707.1 - 4 316
AK299538.1 - 3 452
AK299706.1 - 3 610
AY830839.1 - 5 321

View Full Product Details