Gene Transcripts

Gene Symbol
OGDHL
Entrez Gene ID
55753
Gene Name
oxoglutarate dehydrogenase-like
Gene Aliases
Chromosome Location
Chr. 10: 49734641 - 49762379 on Build GRCh38
UniGene ID
Hs.17860
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018245.2 NP_060715.2 3 387
XM_011539946.1 XP_011538248.1 3 272
XM_011539947.1 XP_011538249.1 3 478
XM_017016401.1 XP_016871890.1 3 351
XM_017016402.1 XP_016871891.1 3 262
XM_017016403.1 XP_016871892.1 2 126
GenBank mRNA AB033116.3 - 2 250
AK001713.1 - 3 331
AK225558.1 - 3 318
AK290226.1 - 2 360
AK307759.1 - 2 146
BC026320.1 - 3 306 4797814
DA238469.1 - 3 387

View Full Product Details