Gene Transcripts

Gene Symbol
KLHL26
Entrez Gene ID
55295
Gene Name
kelch like family member 26
Gene Aliases
Chromosome Location
Chr. 19: 18637028 - 18670492 on Build GRCh38
UniGene ID
Hs.250632
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018316.1 NP_060786.1 3 1711
GenBank mRNA AK001940.1 - 3 1711
AK222656.1 - 3 1711
BC026319.1 - 3 1706 4799495

View Full Product Details