Gene Transcripts
- Gene Symbol
- ANKUB1
- Entrez Gene ID
- 389161
- Gene Name
- ankyrin repeat and ubiquitin domain containing 1
- Gene Aliases
- C3orf16
- Chromosome Location
- Chr. 3: 149760648 - 149792823 on Build GRCh38
- UniGene ID
- Hs.665988
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK098078.1 |
- |
2 |
440 |
|
View Full Product Details