Gene Transcripts

Gene Symbol
TAF8
Entrez Gene ID
129685
Gene Name
TATA-box binding protein associated factor 8
Gene Aliases
II, TAF, TAFII-43, TAFII43, TBN
Chromosome Location
Chr. 6: 42050513 - 42083274 on Build GRCh38
UniGene ID
Hs.520122
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138572.2 NP_612639.2 9 3341
XM_017010241.1 XP_016865730.1 9 1129
XM_017010242.1 XP_016865731.1 9 1292
XM_017010244.1 XP_016865733.1 7 836
XM_017010245.1 XP_016865734.1 7 3039
GenBank mRNA AF086309.1 - 2 587
AL833401.1 - 11 2528

View Full Product Details