Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | XM_005260004.4 | XP_005260061.1 | 7 | 3226 | |
XM_005260005.4 | XP_005260062.1 | 6 | 3055 | ||
XM_005260006.4 | XP_005260063.1 | 7 | 3226 | ||
XM_005260007.4 | XP_005260064.1 | 6 | 3055 | ||
XM_005260008.2 | XP_005260065.1 | 5 | 1070 | ||
XM_005260009.2 | XP_005260066.1 | 3 | 417 | ||
XM_005260010.3 | XP_005260067.1 | 3 | 430 | ||
XM_005260011.2 | XP_005260068.1 | 4 | 899 | ||
XM_005260012.2 | XP_005260069.1 | 3 | 417 | ||
XM_006722828.3 | XP_006722891.1 | 5 | 839 | ||
XM_011528163.2 | XP_011526465.1 | 3 | 491 | ||
GenBank mRNA | AK122890.1 | - | 4 | 446 | |
BC136641.1 | - | 2 | 323 | 9020631 | |
BC144332.1 | - | 2 | 323 | 9052852 |