Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_015656.1 | NP_056471.1 | 15 | 6393 | |
XM_011536641.2 | XP_011534943.1 | 12 | 5538 | ||
GenBank mRNA | AB033062.2 | - | 14 | 6267 | |
AB290161.1 | - | 14 | 6363 | ||
AL117589.1 | - | 4 | 1547 | ||
BC009415.1 | - | 4 | 1581 | ||
BC025353.1 | - | 4 | 2539 | ||
BE540821.1 | - | 1 | 116 |