Gene Transcripts

Gene Symbol
IRGM
Entrez Gene ID
345611
Gene Name
immunity related GTPase M
Gene Aliases
IFI1, IRGM1, LRG-47, LRG47
Chromosome Location
Chr. 5: 150846523 - 150902402 on Build GRCh38
UniGene ID
Hs.519680
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145805.1 NP_001139277.1 2 1128
XM_011537641.2 XP_011535943.1 2 1128
GenBank mRNA BC128168.1 - 1 15
EU742619.1 - 2 1128

View Full Product Details