Gene Transcripts
- Gene Symbol
- C22orf34
- Entrez Gene ID
- 348645
- Gene Name
- chromosome 22 open reading frame 34
- Gene Aliases
- Chromosome Location
- Chr. 22: 49414524 - 49657542 on Build GRCh38
- UniGene ID
- Hs.133159
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_110522.1 |
|
3 |
1416 |
|
NR_110523.1 |
|
5 |
1887 |
|
GenBank mRNA |
BC033837.1 |
- |
5 |
1859 |
|
View Full Product Details