Gene Transcripts

Gene Symbol
C19orf71
Entrez Gene ID
100128569
Gene Name
chromosome 19 open reading frame 71
Gene Aliases
Chromosome Location
Chr. 19: 3539157 - 3544030 on Build GRCh38
UniGene ID
Hs.664978
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135580.1 NP_001129052.1 4 661
XM_011527596.2 XP_011525898.1 4 587
GenBank mRNA BX396051.2 - 4 664

View Full Product Details