Gene Transcripts
- Gene Symbol
- ATP6V1E2
- Entrez Gene ID
- 90423
- Gene Name
- ATPase H+ transporting V1 subunit E2
- Gene Aliases
- ATP6E1, ATP6EL2, ATP6V1EL2, VMA4
- Chromosome Location
- Chr. 2: 46511835 - 46542557 on Build GRCh38
- UniGene ID
- Hs.659656
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001318063.1 |
NP_001304992.1
|
3, 4 |
509 |
|
XM_005264633.2 |
XP_005264690.1
|
2, 3 |
1133 |
|
XM_005264634.2 |
XP_005264691.1
|
2, 3 |
1133 |
|
XM_011533148.2 |
XP_011531450.1
|
2, 3 |
1133 |
|
XM_011533149.2 |
XP_011531451.1
|
2, 3 |
1133 |
|
XM_011533150.2 |
XP_011531452.1
|
3, 4 |
1334 |
|
XM_011533151.2 |
XP_011531453.1
|
2, 3 |
1133 |
|
XM_011533152.2 |
XP_011531454.1
|
2, 3 |
1205 |
|
XM_011533153.2 |
XP_011531455.1
|
2, 3 |
1133 |
|
XM_017005225.1 |
XP_016860714.1
|
3, 4 |
254 |
|
XM_017005226.1 |
XP_016860715.1
|
2, 3 |
1205 |
|
GenBank mRNA |
AL541274.3 |
- |
3, 4 |
509 |
|
- Gene Symbol
- LOC440862
- Entrez Gene ID
- 440862
- Gene Name
- uncharacterized LOC440862
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Hs.437691
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF086285.1 |
- |
2, 3 |
298 |
|
AK123751.1 |
- |
2, 3 |
717 |
|
View Full Product Details