Gene Transcripts
- Gene Symbol
- PRR12
- Entrez Gene ID
- 57479
- Gene Name
- proline rich 12
- Gene Aliases
- KIAA1205
- Chromosome Location
- Chr. 19: 49591177 - 49626439 on Build GRCh38
- UniGene ID
- Hs.590971
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AB033031.1 |
- |
8 |
3132 |
|
BC034003.1 |
- |
6 |
1498 |
|
View Full Product Details