Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001206998.1 | NP_001193927.1 | 3 | 459 | |
NM_032173.3 | NP_115549.2 | 3 | 252 | ||
XM_011530435.2 | XP_011528737.1 | 4 | 448 | ||
XM_011530436.2 | XP_011528738.1 | 5 | 503 | ||
XM_011530437.1 | XP_011528739.1 | 4 | 1173 | ||
XM_011530438.2 | XP_011528740.1 | 4 | 689 | ||
XM_017028990.1 | XP_016884479.1 | 4 | 944 | ||
GenBank mRNA | AB051436.1 | - | 3 | 324 | |
AK096397.1 | - | 3 | 252 | ||
CR456397.1 | - | 3 | 232 |