Gene Transcripts

Gene Symbol
LENG9
Entrez Gene ID
94059
Gene Name
leukocyte receptor cluster member 9
Gene Aliases
Chromosome Location
Chr. 19: 54461796 - 54463711 on Build GRCh38
UniGene ID
Hs.590976
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001301782.1 NP_001288711.1 1 1431
NM_198988.1 NP_945339.2 1 1434
GenBank mRNA AF211976.1 - 1 563
BC015921.1 - 1 1434 4551151

View Full Product Details