Gene Transcripts

Gene Symbol
ACSM2B
Entrez Gene ID
348158
Gene Name
acyl-CoA synthetase medium-chain family member 2B
Gene Aliases
ACSM2, HXMA, HYST1046
Chromosome Location
Chr. 16: 20536261 - 20576379 on Build GRCh38
UniGene ID
Hs.298252
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105069.1 NP_001098539.1 9, 10 1362
NM_182617.3 NP_872423.3 10, 11 1397
XM_005255304.3 XP_005255361.1 9, 10 1301
XM_011545824.2 XP_011544126.1 8, 9 1183
GenBank mRNA AB073604.1 - 10, 11 1328
AK129944.1 - 9, 10 1301
AK289416.1 - 9, 10 1301
AK309601.1 - 9, 10 1362
AY160217.1 - 8, 9 1187

View Full Product Details