Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001320848.1 | NP_001307777.1 | 6 | 907 | |
NM_199336.2 | NP_955368.1 | 5 | 781 | ||
XM_011510745.2 | XP_011509047.1 | 6 | 897 | ||
XM_011510746.2 | XP_011509048.1 | 6 | 1016 | ||
XM_011510747.2 | XP_011509049.1 | 5 | 781 | ||
XM_011510748.2 | XP_011509050.1 | 6 | 897 | ||
XM_017003470.1 | XP_016858959.1 | 6 | 894 | ||
XM_017003471.1 | XP_016858960.1 | 6 | 897 | ||
XM_017003472.1 | XP_016858961.1 | 5 | 830 | ||
GenBank mRNA | AL833869.1 | - | 4 | 581 | |
BC064511.1 | - | 5 | 744 | 6644987 |