Gene Transcripts

Gene Symbol
KRTAP10-2
Entrez Gene ID
386679
Gene Name
keratin associated protein 10-2
Gene Aliases
KAP10.2, KAP18-2, KAP18.2, KRTAP10.2, KRTAP18-2, KRTAP18.2
Chromosome Location
Chr. 21: 44550357 - 44551505 on Build GRCh38
UniGene ID
Hs.664660
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198693.3 NP_941966.1 1 808
GenBank mRNA AB076348.1 - 1 808
AJ566381.1 - 1 109

View Full Product Details