Gene Transcripts

Gene Symbol
OR2T12
Entrez Gene ID
127064
Gene Name
olfactory receptor family 2 subfamily T member 12
Gene Aliases
OR1-57
Chromosome Location
Chr. 1: 248294616 - 248295578 on Build GRCh38
UniGene ID
Hs.553582
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004692.1 NP_001004692.1 1 103

View Full Product Details