Gene Transcripts

Gene Symbol
NANOS2
Entrez Gene ID
339345
Gene Name
nanos C2HC-type zinc finger 2
Gene Aliases
NOS2, ZC2HC12B
Chromosome Location
Chr. 19: 45913214 - 45914778 on Build GRCh38
UniGene ID
Hs.434218
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001029861.2 NP_001025032.1 1 562
GenBank mRNA BC042883.1 - 1 564
BC117484.1 - 1 654 40126035
BC117486.1 - 1 654 40126037

View Full Product Details