Gene Transcripts
- Gene Symbol
- CA10
- Entrez Gene ID
- 56934
- Gene Name
- carbonic anhydrase 10
- Gene Aliases
- CA-RPX, CARPX, HUCEP-15
- Chromosome Location
- Chr. 17: 51630313 - 52160017 on Build GRCh38
- UniGene ID
- Hs.463466
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001082533.1 |
NP_001076002.1
|
3, 4 |
1240 |
|
NM_001082534.1 |
NP_001076003.1
|
3, 4 |
805 |
|
NM_020178.4 |
NP_064563.1
|
2, 3 |
1114 |
|
GenBank mRNA |
AB001597.1 |
- |
2, 3 |
128 |
|
AB036836.1 |
- |
3, 4 |
1030 |
|
AF288385.1 |
- |
3, 4 |
796 |
|
AK313001.1 |
- |
2, 3 |
690 |
|
AL161812.1 |
- |
2, 3 |
130 |
|
AY358509.1 |
- |
2, 3 |
408 |
|
BC020577.1 |
- |
3, 4 |
772 |
4181377 |
BC029865.1 |
- |
3, 4 |
774 |
|
BC047456.1 |
- |
2, 3 |
1027 |
5276935 |
BC068462.1 |
- |
2, 3 |
1073 |
5260321 |
CR749607.1 |
- |
3, 4 |
576 |
|
View Full Product Details