Gene Transcripts

Gene Symbol
Fbxl12
Entrez Gene ID
313782
Gene Name
F-box and leucine-rich repeat protein 12
Gene Aliases
Chromosome Location
Chr. 8: 21656636 - 21661728 on Build Rnor_6.0
UniGene ID
Rn.8828
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025700.1 NP_001020871.1 2 1665
XM_006242639.2 XP_006242701.1 3 2028
GenBank mRNA BC098684.1 - 2 1665

View Full Product Details