Gene Transcripts
- Gene Symbol
- ENY2
- Entrez Gene ID
- 56943
- Gene Name
- ENY2, transcription and export complex 2 subunit
- Gene Aliases
- DC6, Sus1, e(y)2
- Chromosome Location
- Chr. 8: 109334323 - 109345960 on Build GRCh38
- UniGene ID
- Hs.492555
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001193557.1 |
NP_001180486.1
|
3, 4 |
223 |
|
NM_020189.5 |
NP_064574.1
|
3, 4 |
295 |
|
GenBank mRNA |
AF173296.1 |
- |
3, 4 |
365 |
|
AF201940.1 |
- |
3, 4 |
310 |
|
AK000487.1 |
- |
3, 4 |
199 |
|
AK290752.1 |
- |
3, 4 |
239 |
|
AK316560.1 |
- |
3, 4 |
290 |
|
BC007870.2 |
- |
3, 4 |
211 |
4303290 |
BX648621.1 |
- |
1, 2 |
1044 |
|
CR457183.1 |
- |
2, 3 |
149 |
|
DA548665.1 |
- |
3, 4 |
169 |
|
DB168034.1 |
- |
3, 4 |
295 |
|
View Full Product Details