Gene Transcripts

Gene Symbol
Rpo1-3
Entrez Gene ID
360762
Gene Name
RNA polymerase 1-3
Gene Aliases
Chromosome Location
Chr. 12: 9693828 - 9727416 on Build Rnor_6.0
UniGene ID
Rn.6228
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037787.1 NP_001032876.1 3 746
GenBank mRNA BC098735.1 - 3 746
FQ221941.1 - 3 823
FQ232634.1 - 3 805

View Full Product Details