Gene Transcripts

Gene Symbol
Defa-rs1
Entrez Gene ID
13218
Gene Name
defensin, alpha, related sequence 1
Gene Aliases
2010300L12Rik, 2010319H24Rik, CRS1C, CRS1C-2, CRS1C-4, CRS1C-5, Defcr-rs1
Chromosome Location
Chr. 8: 21325888 - 21327020 on Build GRCm38
UniGene ID
Mm.304264
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007844.2 NP_031870.2 2 326
GenBank mRNA AK008584.1 - 2 327
BC117042.1 - 2 301 40126361
BC117044.1 - 2 301 40126363
BC145556.1 - 2 331 9054120
BC171966.1 - 2 331 9054252
EU760893.1 - 1 122
EU760894.1 - 1 122
M33226.1 - 2 306

Gene Symbol
Gm14851
Entrez Gene ID
634825
Gene Name
predicted gene 14851
Gene Aliases
CRS1C-2, OTTMUSG00000018260
Chromosome Location
Chr. 8: 21094971 - 21096050 on Build GRCm38
UniGene ID
Mm.429748
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001177482.1 NP_001170953.1 2 281
GenBank mRNA AV063918.1 - 1 116
AV064248.1 - 1 117
AY761183.1 - 2 281

View Full Product Details