Gene Transcripts

Gene Symbol
CD248
Entrez Gene ID
57124
Gene Name
CD248 molecule
Gene Aliases
CD164L1, TEM1
Chromosome Location
Chr. 11: 66314487 - 66317044 on Build GRCh38
UniGene ID
Hs.195727
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020404.2 NP_065137.1 1 1273
GenBank mRNA AF279142.1 - 1 1261
AJ295846.1 - 1 1273
AK027290.1 - 1 395
BC051340.1 - 1 1260 6339237
BC104484.1 - 1 1273 40007855
BC105633.1 - 1 1273 40007854
KU215671.1 - 1 1211
KU257670.1 - 1 1205

View Full Product Details