Gene Transcripts

Gene Symbol
Psg29
Entrez Gene ID
114872
Gene Name
pregnancy-specific glycoprotein 29
Gene Aliases
AA102956, AA409969, cea17
Chromosome Location
Chr. 7: 17203477 - 17215756 on Build GRCm38
UniGene ID
Mm.322280
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_054064.3 NP_473405.1 5 4180
GenBank mRNA AK141330.1 - 4222
BC114207.1 - 5 4157 30083585

View Full Product Details