Gene Transcripts

Gene Symbol
OR52E4
Entrez Gene ID
390081
Gene Name
olfactory receptor family 52 subfamily E member 4
Gene Aliases
OR11-55
Chromosome Location
Chr. 11: 5884293 - 5885231 on Build GRCh38
UniGene ID
Hs.553739
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005165.1 NP_001005165.1 1 69
GenBank mRNA BC136972.1 - 1 91
BC136974.1 - 1 91 9020966

View Full Product Details