Gene Transcripts

Gene Symbol
Fshb
Entrez Gene ID
25447
Gene Name
follicle stimulating hormone, beta polypeptide
Gene Aliases
Chromosome Location
Chr. 3: 98088321 - 98093144 on Build Rnor_6.0
UniGene ID
Rn.92303
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007597.1 NP_001007598.1 1 82
XM_006234672.2 XP_006234734.1 2 1157
GenBank mRNA BC168724.1 - 2 144
M36804.1 - 1 95

View Full Product Details