Gene Transcripts

Gene Symbol
Vom1r12
Entrez Gene ID
494306
Gene Name
vomeronasal 1 receptor 12
Gene Aliases
V1re23
Chromosome Location
Chr. 1: 60406375 - 60407295 on Build Rnor_6.0
UniGene ID
Rn.129454
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008962.1 NP_001008962.1 1 190
GenBank mRNA AY510364.1 - 1 190

View Full Product Details