Gene Transcripts

Gene Symbol
NMRAL1
Entrez Gene ID
57407
Gene Name
NmrA-like family domain containing 1
Gene Aliases
HSCARG, SDR48A1
Chromosome Location
Chr. 16: 4461677 - 4476320 on Build GRCh38
UniGene ID
Hs.288969
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001305141.1 NP_001292070.1 4 694
NM_001305142.1 NP_001292071.1 4 681
NM_020677.4 NP_065728.1 4 795
XM_006720905.3 XP_006720968.1 5 732
XM_006720906.2 XP_006720969.1 5 687
XM_011522567.2 XP_011520869.1 4 685
XM_017023485.1 XP_016878974.1 3 658
XM_017023486.1 XP_016878975.1 3 661
XM_017023487.1 XP_016878976.1 3 661
XM_017023488.1 XP_016878977.1 4 770
XM_017023489.1 XP_016878978.1 5 783
XM_017023490.1 XP_016878979.1 4 608
XM_017023491.1 XP_016878980.1 3 661
XM_017023492.1 XP_016878981.1 4 770
GenBank mRNA AF225419.1 - 4 800
AK098784.1 - 4 655
BC002927.2 - 4 575 3954035
BC007364.2 - 4 637 3630659

View Full Product Details