Gene Transcripts
- Gene Symbol
- Chchd7
- Entrez Gene ID
- 66433
- Gene Name
- coiled-coil-helix-coiled-coil-helix domain containing 7
- Gene Aliases
- 1810049H20Rik
- Chromosome Location
- Chr. 4: 3938860 - 3951382 on Build GRCm38
- UniGene ID
- Mm.27406
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001190322.2 |
NP_001177251.1
|
3, 4 |
409 |
|
NM_001190323.2 |
NP_001177252.1
|
3, 4 |
285 |
|
NM_001190324.2 |
NP_001177253.1
|
3, 4 |
240 |
|
NM_001285804.1 |
NP_001272733.1
|
3, 4 |
440 |
|
NM_181391.4 |
NP_852056.1
|
3, 4 |
454 |
|
GenBank mRNA |
AA980189.1 |
- |
3, 4 |
237 |
|
AK003308.1 |
- |
3, 4 |
286 |
|
AK007844.1 |
- |
3, 4 |
429 |
|
AK013200.1 |
- |
3, 4 |
427 |
|
AK145105.1 |
- |
3, 4 |
429 |
|
AK168647.1 |
- |
3, 4 |
384 |
|
AK169018.1 |
- |
3, 4 |
429 |
|
BC030366.1 |
- |
3, 4 |
412 |
5370748 |
BX527959.1 |
- |
3, 4 |
201 |
|
CB057699.1 |
- |
3, 4 |
366 |
|
View Full Product Details