Gene Transcripts

Gene Symbol
Rexo4
Entrez Gene ID
227656
Gene Name
REX4, RNA exonuclease 4 homolog (S. cerevisiae)
Gene Aliases
Gm111, Xpmc2h
Chromosome Location
Chr. 2: 26953563 - 26964539 on Build GRCm38
UniGene ID
Mm.291874
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207234.2 NP_997117.2 8 1688
GenBank mRNA AK133982.1 - 8 1608
BC060147.1 - 8 1614 6414260

View Full Product Details