Gene Transcripts

Gene Symbol
ISX
Entrez Gene ID
91464
Gene Name
intestine specific homeobox
Gene Aliases
Pix-1, RAXLX
Chromosome Location
Chr. 22: 35066136 - 35087387 on Build GRCh38
UniGene ID
Hs.567637
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001303508.1 NP_001290437.1 3, 4 1052
GenBank mRNA AB219123.1 - 2, 3 459
AK025181.1 - 2, 3 1341
BC112212.1 - 2, 3 450 8327680
BC113633.1 - 2, 3 438 8322685
CR749372.1 - 3, 4 1053

View Full Product Details