Gene Transcripts

Gene Symbol
GGA1
Entrez Gene ID
26088
Gene Name
golgi associated, gamma adaptin ear containing, ARF binding protein 1
Gene Aliases
Chromosome Location
Chr. 22: 37607235 - 37633564 on Build GRCh38
UniGene ID
Hs.499158
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001560.2 NP_001001560.1 1 401
NM_001172687.1 NP_001166158.1 1 401
NM_013365.4 NP_037497.1 1 401
XM_005261517.3 XP_005261574.1 2 575
XM_011530122.1 XP_011528424.1 2 508
XM_011530124.1 XP_011528426.1 2 575
XM_017028760.1 XP_016884249.1 2 575
XM_017028762.1 XP_016884251.1 2 575
XM_017028763.1 XP_016884252.1 1 41
GenBank mRNA AF190862.1 - 1 29
AF218584.1 - 1 18
AF233521.1 - 1 36
AK075256.1 - 1 379
AK225157.1 - 1 35
AK290548.1 - 1 40
BC010917.1 - 1 45
BC029388.1 - 1 40
BC044629.1 - 1 30 5796877
CR456493.1 - 1 150

View Full Product Details