Gene Transcripts

Gene Symbol
UGT1A9
Entrez Gene ID
54600
Gene Name
UDP glucuronosyltransferase family 1 member A9
Gene Aliases
HLUGP4, LUGP4, UDPGT, UDPGT 1-9, UGT-1I, UGT1-09, UGT1-9, UGT1.9, UGT1A9S, UGT1AI, UGT1I
Chromosome Location
Chr. 2: 233671898 - 233773305 on Build GRCh38
UniGene ID
Hs.554822
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021027.2 NP_066307.1 1 280
GenBank mRNA AF056188.1 - 1 261
AK313510.1 - 1 319
AY435144.1 - 1 243
BC058844.1 - 1 286 4593613

View Full Product Details