Gene Transcripts

Gene Symbol
C6orf47
Entrez Gene ID
57827
Gene Name
chromosome 6 open reading frame 47
Gene Aliases
D6S53E, G4, NG34
Chromosome Location
Chr. 6: 31658298 - 31660772 on Build GRCh38
UniGene ID
Hs.247323
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021184.3 NP_067007.3 1 1774
GenBank mRNA AJ245433.1 - 1 1734
BC012950.1 - 1 1774 3875402

View Full Product Details