Gene Transcripts

Gene Symbol
PELI2
Entrez Gene ID
57161
Gene Name
pellino E3 ubiquitin protein ligase family member 2
Gene Aliases
Chromosome Location
Chr. 14: 56118330 - 56301586 on Build GRCh38
UniGene ID
Hs.657926
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021255.2 NP_067078.1 2 403
XM_005267890.4 XP_005267947.1 2 448
XM_011536990.2 XP_011535292.1 3 113
XM_011536992.2 XP_011535294.1 2 166
XM_017021478.1 XP_016876967.1 2 795
XM_017021479.1 XP_016876968.1 3 9345
GenBank mRNA AF302502.1 - 2 293
AK315727.1 - 2 365
BC009476.2 - 2 403 3942712

View Full Product Details