Gene Transcripts
- Gene Symbol
- EPS8L2
- Entrez Gene ID
- 64787
- Gene Name
- EPS8 like 2
- Gene Aliases
- EPS8R2
- Chromosome Location
- Chr. 11: 705217 - 727727 on Build GRCh38
- UniGene ID
- Hs.55016
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_022772.3 |
NP_073609.2
|
3, 4 |
345 |
|
XM_017018130.1 |
XP_016873619.1
|
4, 5 |
562 |
|
XM_017018131.1 |
XP_016873620.1
|
3, 4 |
433 |
|
XM_017018132.1 |
XP_016873621.1
|
4, 5 |
403 |
|
GenBank mRNA |
AK025588.1 |
- |
3, 4 |
263 |
|
AK025824.1 |
- |
3, 4 |
263 |
|
AK222903.1 |
- |
3, 4 |
258 |
|
AK225311.1 |
- |
3, 4 |
263 |
|
AK294041.1 |
- |
3, 4 |
262 |
|
AY074929.1 |
- |
3, 4 |
370 |
|
BC080636.1 |
- |
3, 4 |
349 |
6572993 |
BC093878.1 |
- |
2, 3 |
138 |
7939723 |
BC101481.1 |
- |
2, 3 |
138 |
8068987 |
BC143242.1 |
- |
2, 3 |
138 |
9051743 |
View Full Product Details