Gene Transcripts

Gene Symbol
RNF38
Entrez Gene ID
152006
Gene Name
ring finger protein 38
Gene Aliases
Chromosome Location
Chr. 9: 36336398 - 36487384 on Build GRCh38
UniGene ID
Hs.333503
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022781.4 NP_073618.3 4 578
NM_194328.2 NP_919309.1 4 745
NM_194329.2 NP_919310.1 3 428
NM_194330.2 NP_919311.1 4 476
NM_194332.2 NP_919313.1 3 595
XM_005251366.3 XP_005251423.1 4 582
XM_005251367.3 XP_005251424.1 4 480
XM_005251368.3 XP_005251425.1 4 594
XM_006716721.3 XP_006716784.1 3 442
XM_011517712.2 XP_011516014.1 5 518
XM_011517713.2 XP_011516015.1 4 582
XM_017014294.1 XP_016869783.1 3 499
XM_017014295.1 XP_016869784.1 4 609
XM_017014296.1 XP_016869785.1 3 179
XM_017014297.1 XP_016869786.1 3 175
GenBank mRNA AF394047.1 - 4 700
AK024996.1 - 2 88
AK093480.1 - 4 644
AK097653.1 - 4 638
AL136817.1 - 4 496
AV690059.2 - 3 319
AX748155.1 - 4 644
BC033786.1 - 3 428 5166325
BG722531.1 - 3 601
BI459742.1 - 3 615
BI520161.1 - 4 474
DA189510.1 - 3 546
DA483270.1 - 3 326

View Full Product Details